siRNA Details
Gene Symbol: SLC25A51P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026540.1 | 1 | 1404 | |||
GenBank mRNA | BC024198.1 | - | 1 | 1405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB524271.1 | - | 1 | 463 | Chr. 6 - 65788820 - 65789306 [+] on Build GRCh38 |
