siRNA Details
Gene Symbol: LINC00242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026781.1 | 2 | 1479 | |||
GenBank mRNA | AK056013.1 | - | 2 | 1479 | ||
BC119778.1 | - | 1 | 1162 | |||
BC122871.1 | - | 1 | 1163 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026781.1 | 2 | 1479 | |||
GenBank mRNA | AK056013.1 | - | 2 | 1479 | ||
BC119778.1 | - | 1 | 1162 | |||
BC122871.1 | - | 1 | 1163 |