siRNA Details
Gene Symbol: C20orf166-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033263.1 | 7 | 3586 | |||
GenBank mRNA | AL832097.1 | - | 1 | 2743 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C20orf166-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033263.1 | 7 | 3586 | |||
GenBank mRNA | AL832097.1 | - | 1 | 2743 |