siRNA Details
Gene Symbol: C20orf166-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033263.1 | 7 | 2090 | |||
GenBank mRNA | AK054875.1 | - | 3 | 2542 | ||
AK056546.1 | - | 7 | 2055 | |||
AL832097.1 | - | 1 | 1246 |
