siRNA Details
Gene Symbol: AGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027032.1 | 2 | 1246 | |||
GenBank mRNA | BC039697.1 | - | 2 | 1165 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027032.1 | 2 | 1246 | |||
GenBank mRNA | BC039697.1 | - | 2 | 1165 |