siRNA Details
Gene Symbol: AGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027032.1 | 2 | 1154 | |||
GenBank mRNA | AK301303.1 | - | 2 | 1128 | ||
BC039697.1 | - | 2 | 1073 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027032.1 | 2 | 1154 | |||
GenBank mRNA | AK301303.1 | - | 2 | 1128 | ||
BC039697.1 | - | 2 | 1073 |