siRNA Details
Gene Symbol: AQP4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026908.1 | 8 | 2230 | |||
GenBank mRNA | AK055069.1 | - | 8 | 2231 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AQP4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026908.1 | 8 | 2230 | |||
GenBank mRNA | AK055069.1 | - | 8 | 2231 |