siRNA Details
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002605.2 | 2 | 897 | |||
NR_109973.1 | 3 | 950 | ||||
GenBank mRNA | AJ412023.1 | - | LIT1929 BCMS |
2 | 895 | |
AJ412024.1 | - | LIT1928 BCMS |
3 | 948 | ||
AK095311.1 | - | 3 | 870 |
