siRNA Details
Gene Symbol: UCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015379.3 | 1 | 187 | |||
GenBank mRNA | AK092401.1 | - | 1 | 173 | ||
BC005351.1 | - | 1 | 39 | |||
BC017567.1 | - | 1 | 43 | |||
BC107722.1 | - | 1 | 43 | |||
DQ343132.1 | - | LIT3336 UCA1 |
1 | 187 | ||
EU334869.1 | - | 1 | 188 | |||
GU357550.1 | - | 1 | 39 | |||
GU799561.1 | - | 1 | 187 | |||
GU799562.1 | - | 1 | 187 | |||
GU799563.1 | - | 1 | 187 | |||
GU799564.1 | - | 1 | 187 | |||
GU799565.1 | - | 1 | 187 | |||
HQ833208.1 | - | 1 | 187 | |||
HQ833209.1 | - | 1 | 187 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX397387.2 | - | 1 | 115 | Chr. 19 - 15829019 - 15835083 [+] on Build GRCh38 |
