siRNA Details
Gene Symbol: PWAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_022009.1 | 1 | 1016 | |||
GenBank mRNA | AF019616.1 | - | LIT2055 PAR1 |
1 | 1020 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PWAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_022009.1 | 1 | 1016 | |||
GenBank mRNA | AF019616.1 | - | LIT2055 PAR1 |
1 | 1020 |