siRNA Details
Gene Symbol: PWAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_022009.1 | 1 | 2216 | |||
GenBank mRNA | AF019616.1 | - | LIT2055 PAR1 |
1 | 2220 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV709519.1 | - | 1 | 77 | Chr. 15 - 25137781 - 25138357 [+] on Build GRCh38 |
