siRNA Details
Gene Symbol: OIP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026757.1 | 4 | 558 | |||
GenBank mRNA | AK172729.1 | - | 4 | 554 | ||
AY007106.1 | - | 4 | 540 | |||
BC037835.1 | - | 4 | 590 | |||
BX647764.1 | - | 4 | 557 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129752.1 | - | 1577 | Chr. - - [-] on Build |
