siRNA Details
Gene Symbol: Nct2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY940663.1 | - | LIT3383 Nct2 |
1 | 153 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nct2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY940663.1 | - | LIT3383 Nct2 |
1 | 153 |