siRNA Details
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
2 | 2012 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
2 | 2012 |