siRNA Details
Gene Symbol: 1810013L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK013883.1 | - | LIT3366 3010002C02 |
1 | 91 | |
AK048963.1 | - | 1 | 468 | |||
AK079605.1 | - | 1 | 220 | |||
AK135924.1 | - | 1 | 3180 | |||
AK163088.1 | - | 1 | 689 |
