siRNA Details
Gene Symbol: 2610100L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033490.1 | 1 | 116 | |||
GenBank mRNA | AK011787.1 | - | LIT3367 2610100L16 |
1 | 91 | |
BE647934.1 | - | 1 | 116 |
