siRNA Details
Gene Symbol: Pinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | DQ059756.1 | - | LIT3461 PINC |
2, 3 | 287 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | DQ059756.1 | - | LIT3461 PINC |
2, 3 | 287 |