siRNA Details
Gene Symbol: 2810037O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK012876.1 | - | LIT3368 2810037O22 |
1 | 1528 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2810037O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK012876.1 | - | LIT3368 2810037O22 |
1 | 1528 |