siRNA Details
Gene Symbol: D630033A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK052706.1 | - | LIT3440 D630033A02Rik |
645 | ||
BE456338.1 | - | 303 | ||||
BE691013.1 | - | 185 |
