siRNA Details
Gene Symbol: D630033A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK052706.1 | - | LIT3440 D630033A02Rik |
566 | ||
BE456338.1 | - | 224 | ||||
BE691013.1 | - | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661268.1 | - | 1 | 566 | Chr. 10 - 122985221 - 122985831 [-] on Build GRCm38 |
