siRNA Details
Gene Symbol: Gm2044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK144366.1 | - | LIT3443 G630041I22Rik |
615 | ||
BB748743.1 | - | 145 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB572889.1 | - | 2 | 617 | Chr. 7 - 139957207 - 139958041 [-] on Build GRCm38 |
