siRNA Details
Gene Symbol: LOC100130927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124783.1 | - | 1 | 3792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AY927635.3 | - | 1 | 1831 | Chr. 6 - 3150019 - 3152205 [+] on Build GRCh38 |
