siRNA Details
Gene Symbol: OR52B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF359417.1 | - | LIT2086 PJCG7 pseudogene |
1 | 417 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR52B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF359417.1 | - | LIT2086 PJCG7 pseudogene |
1 | 417 |