siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400488.1 | - | LIT2213 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2 | 513 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB134898.1 | - | 1 | 38 | Chr. 15 - 25096935 - 25103940 [+] on Build GRCh38 |
