siRNA Details
Gene Symbol: VN1R108P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF253316.1 | - | LIT2078 PHRET pseudogene |
1 | 670 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX091009.1 | - | 2 | 682 | Chr. 20 - 25697076 - 25734972 [+] on Build GRCh38 |
