siRNA Details
Gene Symbol: LOC101928911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110869.1 | 3 | 333 | |||
GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
3 | 334 | |
AY927642.1 | - | 2 | 182 |
