siRNA Details
Gene Symbol: LOC101928911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110869.1 | 1 | 77 | |||
GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
1 | 78 | |
AY927642.1 | - | 1 | 78 |
