siRNA Details
Gene Symbol: PRKXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_073405.1 | 661 | ||||
GenBank mRNA | Y10483.1 | - | LIT2111 PRKXP1 |
1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | Y10482.1 | - | 442 | Chr. - - [-] on Build |
