siRNA Details
Gene Symbol: LOC101929106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110052.1 | 3 | 707 | |||
GenBank mRNA | AK127178.1 | - | 3 | 707 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY329911.1 | - | 1 | 403 | Chr. 3 - 187200020 - 187200464 [-] on Build GRCh38 |
