siRNA Details
Gene Symbol: PRORY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030026.1 | XP_016885515.1 | 1 | 2938 | ||
XM_017030027.1 | XP_016885516.1 | 1 | 2938 | |||
GenBank mRNA | AK026367.1 | - | 1 | 162 |
