siRNA Details
Gene Symbol: PRORY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282471.1 | NP_001269400.1 | 3 | 263 | ||
XM_017030026.1 | XP_016885515.1 | 3 | 5500 | |||
XM_017030027.1 | XP_016885516.1 | 4 | 4955 | |||
GenBank mRNA | AK026367.1 | - | 4 | 462 |
