siRNA Details
Gene Symbol: LINC00639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039982.1 | 3 | 3790 | |||
GenBank mRNA | AK125018.1 | - | 3 | 3785 | ||
AK127318.1 | - | 1 | 2885 | |||
BC035119.1 | - | 4 | 3857 | |||
BC113646.1 | - | 1 | 664 | |||
BC113672.1 | - | 1 | 662 |
