siRNA Details
Gene Symbol: LINC00943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038256.1 | 2 | 236 | |||
GenBank mRNA | AK091521.1 | - | 2 | 709 | ||
AY070435.1 | - | 358 | ||||
BC039096.1 | - | 2 | 218 | |||
DB450910.1 | - | 2 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL538253.3 | - | 1 | 90 | Chr. 12 - 126742736 - 126745093 [+] on Build GRCh38 |
