siRNA Details
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286116.1 | NP_001273045.1 | 9 | 2161 | ||
NM_001286117.1 | NP_001273046.1 | 9 | 2708 | |||
NM_001286118.1 | NP_001273047.1 | 8 | 2284 | |||
NM_001286119.1 | NP_001273048.1 | 9 | 2369 | |||
NM_001286120.1 | NP_001273049.1 | 7 | 2164 | |||
NM_021825.4 | NP_068597.2 | 9 | 2388 | |||
XM_005274156.3 | XP_005274213.1 | 9 | 2375 | |||
XM_005274159.3 | XP_005274216.1 | 10 | 2054 | |||
XM_017018104.1 | XP_016873593.1 | 10 | 2479 | |||
GenBank mRNA | AK026338.1 | - | 1 | 1061 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR604405.1 | - | 1 | 436 | Chr. 11 - 83259299 - 83261158 [-] on Build GRCh38 |
