siRNA Details
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286116.1 | NP_001273045.1 | 9 | 3147 | ||
NM_001286117.1 | NP_001273046.1 | 9 | 3694 | |||
NM_001286118.1 | NP_001273047.1 | 8 | 3270 | |||
NM_001286119.1 | NP_001273048.1 | 9 | 3355 | |||
NM_001286120.1 | NP_001273049.1 | 7 | 3150 | |||
NM_021825.4 | NP_068597.2 | 9 | 3374 |
