siRNA Details
Gene Symbol: MEIOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097747.1 | - | 10 | 2200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK096357.1 | - | 1 | 379 | Chr. 17 - 44689858 - 44693340 [+] on Build GRCh38 |
