siRNA Details
Gene Symbol: DEPDC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017009629.1 | XP_016865118.1 | 8 | 1735 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK023012.1 | - | 1 | 525 | Chr. 5 - 60626720 - 60629009 [-] on Build GRCh38 |
