siRNA Details
Gene Symbol: LOC101928886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110741.1 | 1, 2 | 300 | |||
GenBank mRNA | BC040925.1 | - | 1, 2 | 300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM927328.1 | - | 1, 2 | 299 | Chr. 19 - 56066685 - 56077324 [+] on Build GRCh38 |
