siRNA Details
Gene Symbol: ERV18-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126787.1 | - | 1 | 3180 |
Gene Symbol: UNC13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022223.1 | XP_016877712.1 | 1 | 2318 |
