siRNA Details
Gene Symbol: NTRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720543.3 | XP_006720606.1 | 17 | 14982 | ||
XM_006720544.3 | XP_006720607.1 | 16 | 14958 | |||
XM_017022240.1 | XP_016877729.1 | 18 | 14853 | |||
XM_017022241.1 | XP_016877730.1 | 17 | 14338 | |||
XM_017022243.1 | XP_016877732.1 | 17 | 14334 | |||
XM_017022244.1 | XP_016877733.1 | 17 | 14236 | |||
XM_017022245.1 | XP_016877734.1 | 18 | 14291 | |||
XM_017022246.1 | XP_016877735.1 | 17 | 14392 | |||
XM_017022251.1 | XP_016877740.1 | 10 | 13353 | |||
XM_017022252.1 | XP_016877741.1 | 9 | 13329 | |||
XM_017022253.1 | XP_016877742.1 | 18 | 15024 | |||
XM_017022254.1 | XP_016877743.1 | 18 | 15057 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL109696.1 | - | 1 | 556 | Chr. 15 - 87863523 - 87865601 [-] on Build GRCh38 |
