siRNA Details
Gene Symbol: ZNF444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259034.2 | XP_005259091.1 | 2 | 207 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124209.1 | - | 1 | 163 | Chr. 19 - 56142134 - 56144317 [+] on Build GRCh38 |
