siRNA Details
Gene Symbol: LOC100506985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109781.1 | 3 | 756 | |||
GenBank mRNA | AL557896.3 | - | 3 | 756 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109781.1 | 3 | 756 | |||
GenBank mRNA | AL557896.3 | - | 3 | 756 |