siRNA Details
Gene Symbol: LOC101929109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134278.1 | 1 | 21 | |||
GenBank mRNA | BC043525.1 | - | 1 | 21 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI832418.1 | - | 25 | Chr. - - [-] on Build |
