siRNA Details
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199328.2 | NP_001186257.2 | 5 | 3890 | ||
NM_001199329.2 | NP_001186258.2 | 4 | 3835 | |||
NM_003838.4 | NP_003829.3 | 4 | 4652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126321.1 | - | 1 | 514 | Chr. 1 - 74208119 - 74212064 [+] on Build GRCh38 |
