siRNA Details
Gene Symbol: EIF3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095574.1 | - | 9 | 2691 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK057956.1 | - | 1 | 276 | Chr. 11 - 7997369 - 7999490 [+] on Build GRCh38 |
