siRNA Details
Gene Symbol: LOC93463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135202.1 | 6 | 1184 | |||
GenBank mRNA | AL122100.1 | - | 6 | 1188 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC93463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135202.1 | 6 | 1184 | |||
GenBank mRNA | AL122100.1 | - | 6 | 1188 |