siRNA Details
Gene Symbol: LINC00511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033876.1 | 5 | 1206 | |||
GenBank mRNA | AK129994.1 | - | 6 | 1206 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF215555.1 | - | 108 | Chr. - - [-] on Build |
