siRNA Details
Gene Symbol: LOC101928595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135192.1 | 2 | 1291 | |||
GenBank mRNA | AK095480.1 | - | 2 | 1291 | ||
AK097453.1 | - | 2 | 643 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135192.1 | 2 | 1291 | |||
GenBank mRNA | AK095480.1 | - | 2 | 1291 | ||
AK097453.1 | - | 2 | 643 |