siRNA Details
Gene Symbol: LOC101928595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135192.1 | 1 | 534 | |||
GenBank mRNA | AK095480.1 | - | 1 | 534 | ||
AK097453.1 | - | 2 | 818 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL550229.3 | - | 1 | 500 | Chr. 16 - 30096463 - 30103534 [+] on Build GRCh38 |
