siRNA Details
Gene Symbol: FLJ45513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242791.1 | NP_001229720.1 | 2 | 131 | ||
GenBank mRNA | AK127421.1 | - | 2 | 131 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA386213.1 | - | 2 | 131 | Chr. 17 - 49845910 - 49847303 [+] on Build GRCh38 |
